Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014464.3 | NP_055279.3 | 11 | 1512 | ||
XM_006715062.2 | XP_006715125.1 | 12 | 1495 | |||
XM_011514497.2 | XP_011512799.1 | 9 | 1344 | |||
XM_017010745.1 | XP_016866234.1 | 10 | 1430 | |||
XM_017010746.1 | XP_016866235.1 | 11 | 1270 | |||
XM_017010747.1 | XP_016866236.1 | 9 | 1121 | |||
XM_017010748.1 | XP_016866237.1 | 9 | 950 | |||
GenBank mRNA | AB022277.1 | - | 11 | 1367 | ||
AF195116.1 | - | 11 | 1366 | |||
AF195117.1 | - | 7 | 937 | |||
AK312918.1 | - | 11 | 1476 | |||
AW027949.1 | - | 1 | 289 | |||
BC070278.1 | - | 11 | 1512 | 4607306 |