Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VENTX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014468.3 | NP_055283.1 | 3 | 449 | ||
XM_017016073.1 | XP_016871562.1 | 3 | 444 | |||
GenBank mRNA | AF068006.1 | - | 3 | 449 | ||
BC108915.1 | - | 3 | 449 | 40015551 | ||
BC108916.1 | - | 3 | 449 | 40015553 |