Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114107.4 | NP_001107579.1 | 5 | 685 | ||
NM_001257962.1 | NP_001244891.1 | 5 | 565 | |||
NM_001257963.1 | NP_001244892.1 | 3 | 328 | |||
NM_014476.5 | NP_055291.2 | 6 | 829 | |||
NR_047562.1 | 4 | 497 | ||||
GenBank mRNA | AF002280.1 | - | 6 | 751 | ||
AF002282.1 | - | 5 | 607 | |||
AF039018.1 | - | 6 | 775 | |||
AK129863.1 | - | 4 | 366 | |||
AK222966.1 | - | 6 | 733 | |||
AK313253.1 | - | 6 | 778 | |||
BC027870.1 | - | 5 | 612 | 5205215 | ||
BX647263.1 | - | 2 | 4206 | |||
CD625728.1 | - | 1 | 32 | |||
DA898089.1 | - | 3 | 277 |