Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114107.4 | NP_001107579.1 | 7 | 954 | ||
NM_001257962.1 | NP_001244891.1 | 7 | 834 | |||
NM_001257963.1 | NP_001244892.1 | 5 | 597 | |||
NM_014476.5 | NP_055291.2 | 8 | 1098 | |||
NR_047562.1 | 6 | 766 | ||||
GenBank mRNA | AF002280.1 | - | 8 | 1020 | ||
AF002282.1 | - | 7 | 876 | |||
AF039018.1 | - | 8 | 1044 | |||
AK129863.1 | - | 6 | 635 | |||
AK222966.1 | - | 8 | 1002 | |||
AK313253.1 | - | 8 | 1047 | |||
BC027870.1 | - | 7 | 881 | 5205215 | ||
BX647263.1 | - | 4 | 4475 | |||
CD625728.1 | - | 3 | 301 | |||
CR739525.1 | - | 1 | 55 | |||
DA898089.1 | - | 5 | 546 |