Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286059.1 | NP_001272988.1 | 2 | 419 | ||
NM_001286060.1 | NP_001272989.1 | 2 | 254 | |||
NM_001286061.1 | NP_001272990.1 | 2 | 152 | |||
NM_014488.4 | NP_055303.2 | 3 | 355 | |||
GenBank mRNA | AF498956.1 | - | 1 | 13 | ||
AK291376.1 | - | 2 | 414 | |||
AK310082.1 | - | 1 | 110 | |||
AK312323.1 | - | 2 | 160 | |||
BC014213.2 | - | 3 | 274 | 4763140 | ||
BP310715.1 | - | 2 | 420 | |||
BP336371.1 | - | 2 | 152 | |||
BX641138.1 | - | 2 | 203 | |||
CR542106.1 | - | 1 | 13 | |||
DB151524.1 | - | 2 | 254 | |||
U57092.1 | - | 1 | 31 |