Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014494.2 | NP_055309.2 | 19 | 4816 | ||
XM_005255254.3 | XP_005255311.1 | 18 | 4659 | |||
XM_005255257.4 | XP_005255314.1 | 18 | 4783 | |||
XM_017023143.1 | XP_016878632.1 | 18 | 4760 | |||
XM_017023144.1 | XP_016878633.1 | 17 | 4727 | |||
XM_017023145.1 | XP_016878634.1 | 18 | 4693 | |||
XM_017023146.1 | XP_016878635.1 | 18 | 4825 | |||
XM_017023147.1 | XP_016878636.1 | 18 | 4853 | |||
XM_017023148.1 | XP_016878637.1 | 17 | 4580 | |||
XM_017023149.1 | XP_016878638.1 | 16 | 4577 | |||
XM_017023150.1 | XP_016878639.1 | 17 | 4542 | |||
XM_017023151.1 | XP_016878640.1 | 16 | 4431 | |||
XM_017023152.1 | XP_016878641.1 | 17 | 4332 | |||
XM_017023153.1 | XP_016878642.1 | 18 | 4783 | |||
XM_017023154.1 | XP_016878643.1 | 16 | 4674 | |||
GenBank mRNA | AB040893.1 | - | 14 | 3016 | ||
AJ492221.1 | - | 18 | 4702 | |||
AK001773.1 | - | 2 | 141 | |||
AK025696.1 | - | 12 | 1373 | |||
AL713750.1 | - | 6 | 1569 | |||
AL833757.1 | - | 13 | 1977 | |||
AY035864.1 | - | 16 | 4350 | |||
BC024324.1 | - | 2 | 162 |