Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321727.1 | NP_001308656.1 | 5 | 1637 | ||
NM_001321728.1 | NP_001308657.1 | 5 | 1607 | |||
NM_013247.4 | NP_037379.1 | 5 | 1607 | |||
NM_145074.2 | NP_659540.1 | 4 | 1412 | |||
NR_135769.1 | 5 | 1647 | ||||
NR_135770.1 | 6 | 1075 | ||||
NR_135771.1 | 6 | 1059 | ||||
NR_135772.1 | 6 | 1079 | ||||
GenBank mRNA | AF020760.2 | - | 5 | 1022 | ||
AF141305.1 | - | 5 | 1607 | |||
AF141306.1 | - | 5 | 996 | |||
AF141307.1 | - | 4 | 1068 | |||
AF184911.1 | - | 4 | 827 | |||
AK225932.1 | - | 5 | 1040 | |||
AK291977.1 | - | 5 | 1026 | |||
AK310163.1 | - | 3 | 731 | |||
BC000096.2 | - | 5 | 1033 | 3508944 |