Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001118885.1 | NP_001112357.1 | 10 | 1553 | ||
NM_001118886.1 | NP_001112358.1 | 9 | 1981 | |||
NM_001171942.1 | NP_001165413.1 | 11 | 1720 | |||
NM_002063.3 | NP_002054.1 | 9 | 1981 | |||
XM_006724487.3 | XP_006724550.1 | 10 | 1364 | |||
XM_011545495.2 | XP_011543797.1 | 11 | 2038 | |||
XM_011545496.2 | XP_011543798.1 | 11 | 1748 | |||
XM_017029427.1 | XP_016884916.1 | 10 | 1525 | |||
XM_017029428.1 | XP_016884917.1 | 10 | 1399 | |||
XM_017029429.1 | XP_016884918.1 | 11 | 1944 | |||
GenBank mRNA | AK289561.1 | - | 9 | 1508 | ||
AK297268.1 | - | 9 | 1472 | |||
AK297304.1 | - | 11 | 1657 | |||
AK312591.1 | - | 10 | 1553 | |||
AY437083.1 | - | 9 | 1221 | |||
AY437084.1 | - | 9 | 1221 | |||
BC032864.2 | - | 9 | 1751 | 5263356 | ||
CR450343.1 | - | 9 | 1221 | |||
X52008.1 | - | 9 | 1608 |