Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145076.2 | NP_001138548.1 | 17 | 2139 | ||
NM_019063.4 | NP_061936.2 | 18 | 2313 | |||
XM_005264267.1 | XP_005264324.1 | 19 | 2326 | |||
XM_005264268.3 | XP_005264325.1 | 19 | 2326 | |||
XM_006711991.1 | XP_006712054.1 | 18 | 2152 | |||
XM_006711992.2 | XP_006712055.1 | 20 | 2447 | |||
GenBank mRNA | AF177377.2 | - | 18 | 2275 | ||
AK056880.1 | - | 7 | 815 | |||
AK314702.1 | - | 18 | 2287 | |||
AL832392.1 | - | 4 | 466 | |||
BC140845.1 | - | 18 | 2236 | 9021713 | ||
BC146799.1 | - | 17 | 2113 | 8860304 | ||
BX647731.1 | - | 9 | 1376 |