Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017829.5 | NP_060299.4 | 2, 3 | 289 | ||
NM_033070.2 | NP_149061.1 | 2, 3 | 364 | |||
XM_005261236.3 | XP_005261293.1 | 2, 3 | 290 | |||
XM_011546127.1 | XP_011544429.1 | 2, 3 | 779 | |||
XM_017028781.1 | XP_016884270.1 | 2, 3 | 779 | |||
XM_017028782.1 | XP_016884271.1 | 2, 3 | 342 | |||
XM_017028783.1 | XP_016884272.1 | 2, 3 | 779 | |||
XM_017028784.1 | XP_016884273.1 | 2, 3 | 290 | |||
GenBank mRNA | AF273270.1 | - | 2, 3 | 289 | ||
AF273271.1 | - | 2, 3 | 349 | |||
AK000461.1 | - | 2, 3 | 362 | |||
AK001034.1 | - | 2, 3 | 342 | |||
AK315156.1 | - | 2, 3 | 350 | |||
BC042540.1 | - | 2, 3 | 348 | 4342582 |