Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002067.4 | NP_002058.2 | 4 | 812 | ||
GenBank mRNA | AB209435.1 | - | 4 | 564 | ||
AF011497.1 | - | 4 | 570 | |||
AF493900.1 | - | 4 | 570 | |||
BC089041.1 | - | 4 | 812 | 6085072 | ||
BC096225.1 | - | 4 | 620 | 40005016 | ||
BC096226.3 | - | 4 | 620 | 40005021 | ||
BC096227.1 | - | 4 | 620 | 40005025 | ||
CR457004.1 | - | 4 | 570 | |||
M69013.1 | - | 4 | 756 |