Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282440.1 | NP_001269369.1 | 3 | 657 | ||
NM_001282441.1 | NP_001269370.1 | 5 | 785 | |||
NM_001293092.1 | NP_001280021.1 | 3 | 746 | |||
NM_007353.2 | NP_031379.2 | 4 | 797 | |||
XM_011515288.2 | XP_011513590.1 | 4 | 357 | |||
XM_017012008.1 | XP_016867497.1 | 4 | 680 | |||
GenBank mRNA | AF493901.1 | - | 4 | 633 | ||
AK122830.1 | - | 4 | 569 | |||
AK127842.1 | - | 5 | 785 | |||
AK128701.1 | - | 3 | 657 | |||
AK295830.1 | - | 3 | 437 | |||
BC087537.1 | - | 4 | 810 | 6208783 | ||
BC111464.1 | - | 4 | 801 | 5264798 | ||
L01694.1 | - | 4 | 633 |