Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006496.3 | NP_006487.1 | 4 | 605 | ||
GenBank mRNA | AF493907.1 | - | 4 | 448 | ||
AK311691.1 | - | 5 | 468 | |||
AK312252.1 | - | 4 | 534 | |||
BC025285.1 | - | 4 | 558 | 5433334 | ||
BT019973.1 | - | 4 | 448 | |||
BT019974.1 | - | 4 | 448 | |||
J03005.1 | - | 4 | 456 | |||
J03198.1 | - | 4 | 510 | |||
J03238.1 | - | 4 | 538 | |||
M27543.1 | - | 4 | 490 |