Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297571.1 | NP_001284500.1 | 9 | 1050 | ||
NM_002075.3 | NP_002066.1 | 9 | 1053 | |||
XM_011520953.2 | XP_011519255.1 | 9 | 985 | |||
GenBank mRNA | AB209283.1 | - | 8 | 2194 | ||
AB593098.1 | - | 9 | 979 | |||
AB593099.1 | - | 9 | 978 | |||
AF501884.1 | - | 8 | 843 | |||
AK313091.1 | - | 9 | 930 | |||
BC000115.1 | - | 9 | 966 | 3351520 | ||
BC002454.2 | - | 9 | 973 | 3342882 | ||
BC015920.1 | - | 9 | 803 | 4648399 | ||
BT009800.1 | - | 8 | 843 | |||
M31328.1 | - | 8 | 849 |
Gene Symbol: CDCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092246.1 | - | 5 | 1820 | ||
AK093805.1 | - | 2 | 1917 | |||
AK097957.1 | - | 3 | 1121 |