Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002077.3 | NP_002068.1 | 13 | 1471 | ||
XM_005251929.3 | XP_005251986.1 | 13 | 1403 | |||
XM_006717062.3 | XP_006717125.1 | 13 | 1467 | |||
XM_006717063.3 | XP_006717126.1 | 13 | 1438 | |||
GenBank mRNA | AB208858.1 | - | 8 | 828 | ||
AK302445.1 | - | 13 | 1280 | |||
BC032853.1 | - | 13 | 1441 | 4828793 | ||
U51587.1 | - | 13 | 1463 |