Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039548.2 | NP_001034637.2 | 2 | 899 | ||
XM_011544954.2 | XP_011543256.1 | 3 | 2408 | |||
GenBank mRNA | AK091109.1 | - | 2 | 356 | ||
BC042952.2 | - | 2 | 440 | 5275230 | ||
BC057763.1 | - | 2 | 461 | |||
BC132708.1 | - | 2 | 263 | 40146730 | ||
BC132710.1 | - | 2 | 263 | 40146732 | ||
BI464945.1 | - | 2 | 466 |