Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173595.3 | NP_775866.2 | 16, 17 | 1799 | ||
XM_011538197.2 | XP_011536499.1 | 15, 16 | 1901 | |||
XM_017019183.1 | XP_016874672.1 | 15, 16 | 1982 | |||
XM_017019184.1 | XP_016874673.1 | 16, 17 | 1831 | |||
GenBank mRNA | AB190990.1 | - | 16, 17 | 1816 | ||
AK125379.1 | - | 16, 17 | 1816 | |||
AK126893.1 | - | 9, 10 | 959 |