Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002833.2 | 14 | 1616 | |||
GenBank mRNA | BC066987.1 | - | 14 | 1616 | ||
BX648666.1 | - | 9 | 783 |
Gene Symbol: LOC101928208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005249924.3 | XP_005249981.1 | 1 | 12 |