Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099650.1 | NP_001093120.1 | 6 | 1215 | ||
NM_173601.1 | NP_775872.1 | 7 | 1308 | |||
XM_017019211.1 | XP_016874700.1 | 7 | 1047 | |||
GenBank mRNA | AK125141.1 | - | 6 | 1068 | ||
AL832209.1 | - | 3 | 445 | |||
BC015597.2 | - | 3 | 370 | |||
BC030023.1 | - | 7 | 1316 | 5266607 | ||
BC039145.1 | - | 6 | 1055 | 4508300 |