Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204477.1 | NP_001191406.1 | 4 | 565 | ||
GenBank mRNA | AK093993.1 | - | 2 | 309 | ||
BC029542.1 | - | 4 | 565 | 5298701 | ||
BX379636.2 | - | 2 | 332 |
Gene Symbol: TVP23C-CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204478.1 | NP_001191407.1 | 7 | 976 | ||
NR_037924.1 | 6 | 831 | ||||
GenBank mRNA | AB074181.1 | - | 7 | 906 | ||
AK303475.1 | - | 7 | 945 |