Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM171A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198475.2 | NP_940877.2 | 4, 5 | 762 | ||
XM_017024490.1 | XP_016879979.1 | 2, 3 | 329 | |||
GenBank mRNA | BC029298.1 | - | 4, 5 | 778 | ||
BC030200.1 | - | 4, 5 | 762 |