Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178517.3 | NP_848612.2 | 2 | 445 | ||
XM_005257238.2 | XP_005257295.1 | 2 | 689 | |||
XM_011524646.2 | XP_011522948.1 | 2 | 987 | |||
GenBank mRNA | AB097818.1 | - | 1 | 390 | ||
AK094752.1 | - | 2 | 445 |