Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAEP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024178.1 | 3 | 199 | |||
GenBank mRNA | AK094896.1 | - | 3 | 199 | ||
BC132826.1 | - | 1 | 86 | |||
BC144329.1 | - | 1 | 86 | 9052849 |
Gene Symbol: MRPL45P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033934.1 | 5 | 725 | |||
GenBank mRNA | BC090855.1 | - | 6 | 695 |