Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178171.4 | NP_835465.2 | 7, 8 | 838 | ||
XM_006721832.2 | XP_006721895.1 | 7, 8 | 791 | |||
XM_011524651.2 | XP_011522953.1 | 5, 6 | 554 | |||
XM_017024503.1 | XP_016879992.1 | 7, 8 | 838 | |||
GenBank mRNA | AB093591.1 | - | 6, 7 | 731 | ||
AK096439.1 | - | 8, 9 | 840 | |||
BC109197.2 | - | 8, 9 | 800 | 40005562 |