Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP1-AS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036489.1 | 1 | 256 | |||
GenBank mRNA | AK098180.1 | - | 1 | 225 | ||
DA301744.1 | - | 1 | 258 |