Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046174.2 | 6 | 809 | |||
NR_046454.1 | 5 | 589 | ||||
NR_046455.1 | 4 | 426 | ||||
NR_046456.1 | 7 | 900 | ||||
NR_046457.1 | 6 | 680 | ||||
GenBank mRNA | AB593072.1 | - | 7 | 900 | ||
AB593073.1 | - | 4 | 426 | |||
AB593075.1 | - | 6 | 678 | |||
BC021928.1 | - | 6 | 774 | |||
BC040725.2 | - | 5 | 568 |