Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213602.2 | NP_998767.1 | 6 | 1068 | ||
GenBank mRNA | AK092746.1 | - | 3 | 619 | ||
AK095432.1 | - | 7 | 772 | |||
AK172835.1 | - | 6 | 1068 | |||
AK290400.1 | - | 6 | 922 | |||
AK290402.1 | - | 6 | 932 | |||
AK301189.1 | - | 6 | 949 | |||
AY971516.1 | - | 6 | 1057 |