Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297752.1 | NP_001284681.1 | 5 | 516 | ||
NM_181845.1 | NP_862828.1 | 5 | 444 | |||
XM_005258784.3 | XP_005258841.1 | 5 | 557 | |||
XM_005258785.3 | XP_005258842.1 | 4 | 351 | |||
XM_005258786.3 | XP_005258843.1 | 5 | 516 | |||
XM_017026628.1 | XP_016882117.1 | 6 | 638 | |||
XM_017026629.1 | XP_016882118.1 | 6 | 679 | |||
XM_017026630.1 | XP_016882119.1 | 6 | 642 | |||
XM_017026631.1 | XP_016882120.1 | 6 | 509 | |||
XM_017026632.1 | XP_016882121.1 | 6 | 685 | |||
XM_017026633.1 | XP_016882122.1 | 6 | 634 | |||
XM_017026634.1 | XP_016882123.1 | 5 | 387 | |||
XM_017026635.1 | XP_016882124.1 | 4 | 452 | |||
XM_017026637.1 | XP_016882126.1 | 5 | 557 | |||
XM_017026638.1 | XP_016882127.1 | 4 | 351 | |||
XM_017026639.1 | XP_016882128.1 | 6 | 679 | |||
GenBank mRNA | AK098175.1 | - | 5 | 434 | ||
AK294852.1 | - | 5 | 473 | |||
BC045755.2 | - | 3 | 198 | |||
BE261849.1 | - | 5 | 413 | |||
DC324604.1 | - | 5 | 444 | |||
DC384756.1 | - | 5 | 372 |