Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297752.1 | NP_001284681.1 | 6 | 675 | ||
NM_181845.1 | NP_862828.1 | 7 | 730 | |||
XM_005258784.3 | XP_005258841.1 | 7 | 843 | |||
XM_005258785.3 | XP_005258842.1 | 6 | 637 | |||
XM_005258786.3 | XP_005258843.1 | 7 | 802 | |||
XM_017026628.1 | XP_016882117.1 | 8 | 924 | |||
XM_017026629.1 | XP_016882118.1 | 8 | 965 | |||
XM_017026630.1 | XP_016882119.1 | 8 | 928 | |||
XM_017026631.1 | XP_016882120.1 | 8 | 795 | |||
XM_017026632.1 | XP_016882121.1 | 8 | 971 | |||
XM_017026633.1 | XP_016882122.1 | 8 | 920 | |||
XM_017026634.1 | XP_016882123.1 | 7 | 673 | |||
XM_017026635.1 | XP_016882124.1 | 6 | 738 | |||
XM_017026636.1 | XP_016882125.1 | 2 | 6443 | |||
XM_017026637.1 | XP_016882126.1 | 6 | 716 | |||
XM_017026638.1 | XP_016882127.1 | 5 | 510 | |||
XM_017026639.1 | XP_016882128.1 | 7 | 838 | |||
GenBank mRNA | AK098175.1 | - | 6 | 587 | ||
AK294852.1 | - | 7 | 753 | |||
AY166784.1 | - | 2 | 211 |