Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF615 SIRNA Assays ›
Gene Symbol:
ZNF615
Gene Name:
zinc finger protein 615
Gene Aliases:
-
Chromosome Location:
Chr.19: 51991332 - 52008230 on Build GRCh38
UniGene ID:
Hs.368355
Species:
Human
siRNA ID
HSS178687
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF615
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199324.1
NP_001186253.1
7
1589
NM_001321317.1
NP_001308246.1
4
1314
NM_001321318.1
NP_001308247.1
6
1385
NM_001321319.1
NP_001308248.1
6
1551
NM_001321320.1
NP_001308249.1
5
1518
NM_001321321.1
NP_001308250.1
6
1544
NM_001321322.1
NP_001308251.1
6
1539
NM_001321323.1
NP_001308252.1
5
1347
NM_198480.3
NP_940882.3
6
1556
XM_011526824.1
XP_011525126.1
8
1867
XM_011526825.1
XP_011525127.1
8
1993
XM_011526826.1
XP_011525128.1
7
1833
XM_011526829.1
XP_011525131.1
6
1602
XM_011526830.1
XP_011525132.1
5
1405
XM_017026648.1
XP_016882137.1
7
1834
XM_017026649.1
XP_016882138.1
7
1960
XM_017026650.1
XP_016882139.1
7
1667
XM_017026651.1
XP_016882140.1
8
2080
GenBank mRNA
AK091029.1
-
1
43
AK096691.1
-
1
715
AK128361.1
-
6
1355
AK131494.1
-
6
1531
AK291734.1
-
7
1562
AK294977.1
-
4
1314
BC053884.1
-
6
1624
BC105100.1
-
4
1277
8144103
BC105102.1
-
4
1277
8144105
BC112961.1
-
6
1498
BC143429.1
-
5
1310
9051937
BX647556.1
-
6
1517
BX648594.1
-
1
294
CR936612.1
-
7
1569
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline