Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288791.1 | NP_001275720.1 | 9 | 657 | ||
NM_001288792.1 | NP_001275721.1 | 8 | 773 | |||
NM_001288793.1 | NP_001275722.1 | 6 | 506 | |||
NM_198481.3 | NP_940883.2 | 9 | 866 | |||
NR_110142.1 | 10 | 955 | ||||
XM_011526845.1 | XP_011525147.1 | 10 | 1263 | |||
XM_011526846.1 | XP_011525148.1 | 10 | 1173 | |||
XM_011526847.1 | XP_011525149.1 | 11 | 1241 | |||
XM_011526848.1 | XP_011525150.1 | 9 | 1080 | |||
XM_011526849.1 | XP_011525151.1 | 9 | 969 | |||
GenBank mRNA | AY358542.1 | - | 9 | 875 | ||
BC100943.2 | - | 8 | 750 | 40004095 | ||
BG189223.1 | - | 5 | 305 | |||
BQ446114.1 | - | 1 | 163 | |||
DQ479397.1 | - | 9 | 707 | |||
FJ584316.1 | - | 9 | 701 | |||
FJ584317.1 | - | 9 | 701 | |||
FJ584318.1 | - | 8 | 608 | |||
FJ584319.1 | - | 6 | 341 | |||
FJ584320.1 | - | 9 | 492 | |||
FJ882051.1 | - | 10 | 790 | |||
FN398145.1 | - | 9 | 694 |