Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HKR1 SIRNA Assays ›
Gene Symbol:
HKR1
Gene Name:
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases:
ZNF875
Chromosome Location:
Chr.19: 37317900 - 37364455 on Build GRCh38
UniGene ID:
Hs.657027
Species:
Human
siRNA ID
HSS178695
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HKR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181786.2
NP_861451.1
6
760
XM_017026672.1
XP_016882161.1
4
948
XM_017026673.1
XP_016882162.1
4
1014
XM_017026674.1
XP_016882163.1
8
1314
XM_017026675.1
XP_016882164.1
8
1311
XM_017026676.1
XP_016882165.1
5
609
XM_017026677.1
XP_016882166.1
5
596
XM_017026678.1
XP_016882167.1
5
606
XM_017026679.1
XP_016882168.1
5
593
XM_017026680.1
XP_016882169.1
8
976
XM_017026681.1
XP_016882170.1
7
924
XM_017026682.1
XP_016882171.1
8
973
XM_017026683.1
XP_016882172.1
7
835
XM_017026684.1
XP_016882173.1
6
697
XM_017026685.1
XP_016882174.1
4
542
XM_017026686.1
XP_016882175.1
7
832
XM_017026687.1
XP_016882176.1
6
694
XM_017026688.1
XP_016882177.1
4
2766
XM_017026689.1
XP_016882178.1
6
780
XM_017026690.1
XP_016882179.1
4
560
XM_017026691.1
XP_016882180.1
3
340
XM_017026692.1
XP_016882181.1
8
910
XM_017026693.1
XP_016882182.1
9
1031
XM_017026694.1
XP_016882183.1
9
981
XM_017026695.1
XP_016882184.1
8
953
XM_017026696.1
XP_016882185.1
7
805
XM_017026697.1
XP_016882186.1
6
727
XM_017026698.1
XP_016882187.1
6
705
XM_017026699.1
XP_016882188.1
4
3541
XM_017026700.1
XP_016882189.1
6
706
XM_017026701.1
XP_016882190.1
8
904
XM_017026702.1
XP_016882191.1
7
826
XM_017026703.1
XP_016882192.1
5
603
XM_017026704.1
XP_016882193.1
5
665
XM_017026705.1
XP_016882194.1
2
662
XM_017026707.1
XP_016882196.1
5
1078
XM_017026708.1
XP_016882197.1
5
1081
GenBank mRNA
AB013897.1
-
5
607
AK056243.1
-
6
694
AK128116.1
-
9
1313
AK223616.1
-
5
600
AK299969.1
-
5
574
AK301601.1
-
4
535
AK307852.1
-
1
319
BC017256.2
-
6
760
3356680
BC053845.1
-
4
597
5493685
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline