Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM19A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004440.1 | NP_001004440.1 | 4 | 503 | ||
NM_182759.2 | NP_877436.1 | 4 | 435 | |||
XM_005270766.3 | XP_005270823.1 | 5 | 722 | |||
XM_006710582.3 | XP_006710645.1 | 5 | 790 | |||
GenBank mRNA | AY325116.1 | - | 4 | 435 | ||
AY325119.1 | - | 4 | 503 | |||
BC112268.1 | - | 4 | 497 | 8327736 | ||
BC117479.1 | - | 4 | 497 | 40126030 | ||
BC143949.1 | - | 4 | 466 | |||
BC144042.1 | - | 4 | 486 | |||
BC144043.1 | - | 4 | 429 | 9052563 |