Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010881.1 | NP_001010881.1 | 12 | 2807 | ||
XM_011541269.2 | XP_011539571.2 | 13 | 3054 | |||
XM_011541271.2 | XP_011539573.2 | 13 | 2849 | |||
XM_011541272.2 | XP_011539574.1 | 13 | 2903 | |||
XM_011541274.2 | XP_011539576.2 | 12 | 2921 | |||
XM_011541275.2 | XP_011539577.2 | 12 | 2718 | |||
XM_011541276.2 | XP_011539578.1 | 13 | 2903 | |||
XM_011541277.2 | XP_011539579.1 | 13 | 2903 | |||
XM_011541278.2 | XP_011539580.1 | 13 | 2903 | |||
XM_011541280.2 | XP_011539582.2 | 8 | 1154 | |||
XM_011541281.2 | XP_011539583.2 | 8 | 1135 | |||
XM_017001062.1 | XP_016856551.1 | 14 | 3041 | |||
XM_017001063.1 | XP_016856552.1 | 13 | 2894 | |||
XM_017001064.1 | XP_016856553.1 | 13 | 2903 | |||
GenBank mRNA | AL833926.1 | - | 1 | 123 | ||
AL834308.1 | - | 5 | 774 |