Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320185.1 | NP_001307114.1 | 4 | 278 | ||
NM_182975.3 | NP_892020.1 | 4 | 278 | |||
NM_182976.3 | NP_892021.1 | 4 | 278 | |||
XM_005270779.4 | XP_005270836.1 | 4 | 258 | |||
XM_011541290.2 | XP_011539592.1 | 4 | 273 | |||
GenBank mRNA | AK000410.1 | - | 4 | 245 | ||
AK297073.1 | - | 4 | 203 | |||
BC014899.1 | - | 4 | 236 | |||
BC052645.1 | - | 4 | 252 | |||
BC053888.1 | - | 4 | 239 | |||
BC070341.1 | - | 4 | 222 | |||
BC090866.1 | - | 4 | 272 | 6503526 | ||
CN299618.1 | - | 4 | 288 | |||
DB111509.1 | - | 4 | 254 |