Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080539.1 | NP_001074008.1 | 20, 21 | 2392 | ||
XM_006712438.3 | XP_006712501.1 | 19, 20 | 2329 | |||
XM_006712439.3 | XP_006712502.1 | 4, 5 | 976 | |||
XM_011510985.2 | XP_011509287.1 | 18, 19 | 2121 | |||
XM_011510987.2 | XP_011509289.1 | 12, 13 | 1312 | |||
XM_011510988.2 | XP_011509290.1 | 16, 17 | 1838 | |||
XM_017003861.1 | XP_016859350.1 | 18, 19 | 2236 | |||
XM_017003862.1 | XP_016859351.1 | 18, 19 | 2215 | |||
XM_017003863.1 | XP_016859352.1 | 19, 20 | 2329 | |||
XM_017003864.1 | XP_016859353.1 | 19, 20 | 2329 | |||
XM_017003865.1 | XP_016859354.1 | 16, 17 | 1853 | |||
XM_017003866.1 | XP_016859355.1 | 13, 14 | 1528 | |||
GenBank mRNA | AK096130.1 | - | 2, 3 | 480 | ||
AK125768.1 | - | 5, 6 | 955 | |||
AK301665.1 | - | 5, 6 | 580 | |||
AK302685.1 | - | 15, 16 | 1760 | |||
AL834537.1 | - | 4, 5 | 871 | |||
BC062449.1 | - | 2, 3 | 472 | |||
BC062658.1 | - | 4, 5 | 932 |