Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173647.3 | NP_775918.2 | 6 | 1157 | ||
XM_005263920.2 | XP_005263977.1 | 6 | 1157 | |||
XM_005263921.3 | XP_005263978.1 | 6 | 1157 | |||
XM_011510990.2 | XP_011509292.1 | 6 | 828 | |||
GenBank mRNA | AK074985.1 | - | 6 | 1131 | ||
AK075141.1 | - | 6 | 1131 | |||
AK129799.1 | - | 6 | 1142 | |||
AY450390.1 | - | 6 | 1131 | |||
BC019355.1 | - | 6 | 1108 | 3956746 | ||
BC045743.1 | - | 6 | 1131 | 5260812 |