Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173648.3 | NP_775919.3 | 23 | 4334 | ||
XM_011510991.2 | XP_011509293.1 | 23 | 4409 | |||
XM_011510992.2 | XP_011509294.1 | 23 | 4409 | |||
XM_011510993.2 | XP_011509295.1 | 22 | 4229 | |||
XM_017003867.1 | XP_016859356.1 | 23 | 4406 | |||
XM_017003868.1 | XP_016859357.1 | 23 | 4408 | |||
XM_017003869.1 | XP_016859358.1 | 23 | 4407 | |||
XM_017003870.1 | XP_016859359.1 | 17 | 3231 | |||
XM_017003871.1 | XP_016859360.1 | 12 | 2546 | |||
GenBank mRNA | AK096821.1 | - | 2 | 868 | ||
AK129847.1 | - | 13 | 2548 | |||
BC115378.2 | - | 13 | 2539 | 40028485 | ||
BX649042.1 | - | 12 | 1936 |