Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178145.1 | NP_001171616.1 | 2 | 1270 | ||
NM_001178146.1 | NP_001171617.1 | 2 | 1117 | |||
NM_178822.4 | NP_849144.2 | 6 | 7017 | |||
XM_011512708.2 | XP_011511010.1 | 12 | 7826 | |||
XM_011512709.2 | XP_011511011.1 | 11 | 7557 | |||
GenBank mRNA | AK093069.1 | - | 2 | 1253 | ||
AK098838.1 | - | 1 | 460 | |||
AK128160.1 | - | 3 | 3066 | |||
AY273815.1 | - | 6 | 7017 | |||
BC031063.1 | - | 2 | 1107 | |||
BC045734.1 | - | 2 | 1108 |