Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198281.2 | NP_938022.2 | 2 | 1429 | ||
XM_005262936.3 | XP_005262993.1 | 2 | 1669 | |||
XM_005262937.4 | XP_005262994.1 | 2 | 1723 | |||
XM_017008043.1 | XP_016863532.1 | 2 | 3506 | |||
XM_017008044.1 | XP_016863533.1 | 2 | 1227 | |||
GenBank mRNA | AB095947.2 | - | 1 | 1994 | ||
AK124616.1 | - | 2 | 1212 | |||
BC126371.1 | - | 1 | 1043 | 8992087 |