Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173829.3 | NP_776190.1 | 3 | 317 | ||
XM_011543344.2 | XP_011541646.1 | 2 | 449 | |||
GenBank mRNA | AK094073.1 | - | 317 | |||
AL832348.1 | - | 3 | 274 | |||
BC047006.1 | - | 3 | 246 | |||
BC108716.1 | - | 3 | 247 | 6184559 | ||
BC127085.1 | - | 3 | 255 | 40122236 | ||
BC127086.1 | - | 3 | 255 | 40122239 |