Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173680.3 | NP_775951.2 | 3 | 232 | ||
GenBank mRNA | AW402889.1 | - | 1 | 50 | ||
BC015152.2 | - | 2 | 170 | 3546283 | ||
DB472309.1 | - | 3 | 234 |
Gene Symbol: REPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL046625.1 | - | 3 | 308 |