Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173685.2 | NP_775956.1 | 6 | 690 | ||
XM_005250875.2 | XP_005250932.1 | 6 | 698 | |||
XM_005250876.4 | XP_005250933.1 | 5 | 591 | |||
XM_011516974.2 | XP_011515276.1 | 7 | 760 | |||
XM_011516975.1 | XP_011515277.1 | 6 | 655 | |||
XM_017013330.1 | XP_016868819.1 | 4 | 871 | |||
XM_017013331.1 | XP_016868820.1 | 7 | 728 | |||
XM_017013332.1 | XP_016868821.1 | 4 | 889 | |||
XM_017013333.1 | XP_016868822.1 | 7 | 800 | |||
GenBank mRNA | AK057002.1 | - | 6 | 660 | ||
BC032797.1 | - | 6 | 689 | 5109691 |