Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM78A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033387.3 | NP_203745.2 | 2 | 874 | ||
XM_011518567.1 | XP_011516869.1 | 3 | 417 | |||
XM_011518568.2 | XP_011516870.1 | 2 | 1120 | |||
GenBank mRNA | BC029924.2 | - | 2 | 288 |