Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRB10 SIRNA Assays ›
Gene Symbol:
GRB10
Gene Name:
growth factor receptor bound protein 10
Gene Aliases:
GRB-IR, Grb-10, IRBP, MEG1, RSS
Chromosome Location:
Chr.7: 50590063 - 50793462 on Build GRCh38
UniGene ID:
Hs.164060
Species:
Human
siRNA ID
HSS178911
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRB10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001549.2
NP_001001549.1
6
749
NM_001001550.2
NP_001001550.1
6
829
NM_001001555.2
NP_001001555.1
7
1076
NM_005311.4
NP_005302.3
6
749
XM_011515304.1
XP_011513606.1
9
1188
XM_011515312.2
XP_011513614.1
8
1476
XM_011515316.1
XP_011513618.1
7
1021
XM_011515323.2
XP_011513625.1
4
2525
XM_017012029.1
XP_016867518.1
8
1408
XM_017012030.1
XP_016867519.1
7
1342
XM_017012031.1
XP_016867520.1
7
1273
XM_017012032.1
XP_016867521.1
13
1785
XM_017012033.1
XP_016867522.1
10
1323
XM_017012034.1
XP_016867523.1
8
924
XM_017012035.1
XP_016867524.1
14
2834
XM_017012036.1
XP_016867525.1
15
2619
XM_017012037.1
XP_016867526.1
15
2456
XM_017012038.1
XP_016867527.1
14
2406
XM_017012039.1
XP_016867528.1
11
1653
XM_017012040.1
XP_016867529.1
11
2189
XM_017012041.1
XP_016867530.1
13
2310
XM_017012042.1
XP_016867531.1
9
1230
XM_017012043.1
XP_016867532.1
10
1420
XM_017012044.1
XP_016867533.1
6
1019
XM_017012045.1
XP_016867534.1
6
683
XM_017012046.1
XP_016867535.1
10
2035
XM_017012047.1
XP_016867536.1
15
3018
XM_017012048.1
XP_016867537.1
14
2902
XM_017012049.1
XP_016867538.1
15
2524
XM_017012050.1
XP_016867539.1
14
2474
XM_017012051.1
XP_016867540.1
13
2378
XM_017012052.1
XP_016867541.1
13
2718
XM_017012053.1
XP_016867542.1
14
2340
XM_017012054.1
XP_016867543.1
13
2290
XM_017012055.1
XP_016867544.1
10
2073
XM_017012056.1
XP_016867545.1
9
2905
XM_017012057.1
XP_016867546.1
12
2194
XM_017012058.1
XP_016867547.1
8
1065
XM_017012059.1
XP_016867548.1
8
1187
XM_017012060.1
XP_016867549.1
16
3151
XM_017012061.1
XP_016867550.1
7
1017
XM_017012062.1
XP_016867551.1
6
778
XM_017012063.1
XP_016867552.1
14
2883
XM_017012064.1
XP_016867553.1
13
2767
XM_017012065.1
XP_016867554.1
11
2219
XM_017012066.1
XP_016867555.1
9
2578
XM_017012067.1
XP_016867556.1
9
1081
XM_017012068.1
XP_016867557.1
9
1947
GenBank mRNA
AB000731.1
-
6
724
AF000017.1
-
6
718
AF001534.1
-
6
749
AJ271366.1
-
9
1188
AK289511.1
-
8
1043
BC024285.1
-
7
853
4590330
BX649028.1
-
6
809
D86962.1
-
8
1481
U34355.1
-
6
749
U66065.1
-
6
829
U69276.1
-
6
831
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of Wnt signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
negative regulation of phosphorylation
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0