Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000828.4 | NP_000819.3 | 12 | 2223 | ||
NM_007325.4 | NP_015564.4 | 12 | 2223 | |||
GenBank mRNA | BC117464.1 | - | 12 | 2095 | 40126015 | |
U10301.1 | - | 12 | 1983 | |||
U10302.1 | - | 12 | 1983 |