Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000828.4 | NP_000819.3 | 3 | 743 | ||
NM_007325.4 | NP_015564.4 | 3 | 743 | |||
GenBank mRNA | AK297136.1 | - | 3 | 429 | ||
BC117464.1 | - | 3 | 615 | 40126015 | ||
U10301.1 | - | 3 | 503 | |||
U10302.1 | - | 3 | 503 |