Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085454.1 | NP_001078923.1 | 19 | 2206 | ||
NM_014030.3 | NP_054749.2 | 18 | 2179 | |||
XM_011524684.1 | XP_011522986.1 | 19 | 2047 | |||
XM_011524685.1 | XP_011522987.1 | 18 | 2020 | |||
GenBank mRNA | AB209537.1 | - | 19 | 2005 | ||
AF124490.1 | - | 18 | 1979 | |||
AK000863.1 | - | 2 | 206 | |||
AK024826.1 | - | 13 | 1331 | |||
AK294785.1 | - | 20 | 2084 | |||
AK297443.1 | - | 16 | 1648 | |||
AK299615.1 | - | 10 | 1076 | |||
AK299932.1 | - | 19 | 1981 | |||
BC006227.2 | - | 2 | 213 | |||
BC018998.2 | - | 1 | 121 | |||
BC032098.1 | - | 1 | 130 | |||
BC067358.1 | - | 18 | 2179 | |||
BX537553.1 | - | 10 | 1914 | |||
BX537554.1 | - | 10 | 1914 | |||
BX648097.1 | - | 10 | 1916 |