Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014049.4 | NP_054768.2 | 3 | 493 | ||
NR_033426.1 | 3 | 669 | ||||
XM_017006264.1 | XP_016861753.1 | 3 | 657 | |||
GenBank mRNA | AB209427.1 | - | 3 | 2186 | ||
AF327351.1 | - | 3 | 323 | |||
AK022568.1 | - | 2 | 419 | |||
AK024012.1 | - | 3 | 337 | |||
AK130311.1 | - | 3 | 364 | |||
AK294597.1 | - | 3 | 324 | |||
AK297952.1 | - | 3 | 537 | |||
AK310644.1 | - | 2 | 266 | |||
BC007970.2 | - | 3 | 370 | 4304209 | ||
BC013354.2 | - | 3 | 347 | 3935925 |