Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142302.1 | NP_001135774.1 | 2 | 257 | ||
NM_014157.3 | NP_054876.2 | 2 | 257 | |||
XM_011523038.2 | XP_011521340.1 | 2 | 112 | |||
XM_011523039.2 | XP_011521341.1 | 4 | 1005 | |||
XM_011523040.2 | XP_011521342.1 | 2 | 530 | |||
XM_011523041.2 | XP_011521343.1 | 2 | 98 | |||
XM_011523042.2 | XP_011521344.1 | 2 | 257 | |||
XM_017023176.1 | XP_016878665.1 | 3 | 420 | |||
GenBank mRNA | AF161550.1 | - | 2 | 216 | ||
AK299066.1 | - | 2 | 257 | |||
AK314300.1 | - | 2 | 246 | |||
AL136785.1 | - | 2 | 234 | |||
BC126445.1 | - | 2 | 226 | 8992161 | ||
BC126447.1 | - | 2 | 226 | 8992163 | ||
CR533506.1 | - | 2 | 178 | |||
DC369542.1 | - | 2 | 258 |